MilliporeSigma™ Supelco™ Empty Glass GC Column, for Agilent 5880, 5890, 6890, and 7890 (Configuration B)
Silane treated empty glass column. Fittings are not included.
Manufacturer: MilliporeSigma™ Supelco™ 21642
Description
X (length of injector arm) = 11.02 in. (280 mm). Y (length of detector arm) = 7.09 in. (180 mm). S (span, injector to detector) = 9.0 in. (229 mm). This column fits Agilent 5880, 5890, 6890, and 7890 (configuration B, on-column injection, TCD only).Specifications
Empty | |
Gas Chromatography (GC) | |
Glass Column |
2 mm | |
3 m |
Agilent is a registered trademark of Agilent Technologies, Inc.