Please login to your online account to display your discounted pricing

EMPA, Tocris™

Highly potent and selective OX2 antagonist; brain penetrant

$266.69 - $1,121.62

Chemical Identifiers

Chemical Name or Material EMPA
Assay Percent Range >98%
Molecular Formula C23H26N4O4S
Formula Weight 454.54
Synonym N-Ethyl-2-[(6-methoxy-3-pyridinyl)[(2-methylphenyl)sulfonyl]amino]-N-(3-pyridinylmethyl)-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

455810

 
r & d systems
RDS455810
10mg Each for $266.69

455850

 
r & d systems
RDS455850
50mg Each for $1,121.62
Description & Specifications

Specifications

Formula Weight 454.54
Quantity 10mg
Chemical Name or Material EMPA
Recommended Storage Store at -20°C
Synonym N-Ethyl-2-[(6-methoxy-3-pyridinyl)[(2-methylphenyl)sulfonyl]amino]-N-(3-pyridinylmethyl)-acetamide
Molecular Formula C23H26N4O4S
Assay Percent Range >98%
CAS 680590-49-2
Safety and Handling

Store at -20°C