Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Emerin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Emerin Ab-1, Mouse Monoclonal Antibody.

$203.31 - $801.53

Specifications

Antigen Emerin Ab-1
Clone 4G5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1751R7

 
thermo scientific™ lab vision™
MS1751R7
7mL Each for $499.73

MS1751S

 
thermo scientific™ lab vision™
MS1751S
1mL Each for $801.53

MS1751S0

 
thermo scientific™ lab vision™
MS1751S0
100µL Each for $203.31

MS1751S1

 
thermo scientific™ lab vision™
MS1751S1
500µL Each for $589.65
Description & Specifications

Specifications

Antigen Emerin Ab-1
Clone 4G5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Immunogen Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ Emerin Ab-1, Mouse Monoclonal Antibody.

Emerin is a serine-rich, ubiquitously expressed protein with the highest concentration in skeletal and cardiac muscles. It is localized in the nuclear membrane and is coded by the STA gene at the  Xq28 locus. Emerin deficiency is the main causal feature of Emery-Dreifuss muscular dystrophy (EDMD), which is an X-linked disorder leading to weakening of muscles and cardiomyopathy presented as heart block.

Host Species: Mouse

Clone: 4G5

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Near N-terminus

Immunogen: Recombinant protein corresponding to 222 amino acid fragment near N-terminus of human emerin protein

Molecular Weight: 34 kDa

Positive Control: Tonsil

Cellular Localization: Perinuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO