Please login to your online account to display your discounted pricing

EMDT oxalate, Tocris™

Selective 5-HT6 agonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material EMDT oxalate
Assay Percent Range >98%
Molecular Formula C15H22N2O·C2H2O4
Formula Weight 336.38
Synonym 2-Ethyl-5-methoxy-N, N-dimethyl-1H-indole-3-ethanamine oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

420910

 
r & d systems
420910
10mg Each for $201.36

420950

 
r & d systems
420950
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 336.38
Quantity 10mg
Chemical Name or Material EMDT oxalate
CAS 263744-72-5
Synonym 2-Ethyl-5-methoxy-N, N-dimethyl-1H-indole-3-ethanamine oxalate
Assay Percent Range >98%
Molecular Formula C15H22N2O·C2H2O4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C