missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore ZHE Hazardous Waste Filtration System Replacement Parts and Accessories: 90mm Diameter

Designated by the US EPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP)

Manufacturer:  EMD Millipore YY2209068

 View more versions of this product

Catalog No. YY2209068


 

Specifications Description & Specifications

Specifications

O-ring, 2-235, 90mm, fluoroelastomer
Fluoroelastomer
ZHE Hazardous Waste Filtration System (90 mm)