missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore ZHE Hazardous Waste Filtration System Replacement Parts and Accessories: 90mm Diameter

Designated by the US EPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP)

Manufacturer:  EMD Millipore YY2209064

 View more versions of this product

Catalog No. YY2209064


 

Specifications Description & Specifications

Specifications

Support Screen 90mm
Stainless Steel
ZHE Hazardous Waste Filtration System (90 mm)