Please login to your online account to display your discounted pricing

EMD Millipore ZHE Hazardous Waste Filtration System Replacement Parts and Accessories: 90mm Diameter


Designated by the US EPA as a suitable apparatus for Toxicity Characteristic Leaching Procedure (TCLP)

Manufacturer: emd millipore  YT3009015

 View more versions of this product

Catalog No. YT3009015

  • / Each
 

Description & Specifications

Specifications

Description ZHE Tight Syringe, 50mL
For Use With (Equipment) ZHE Hazardous Waste Filtration System (90 mm)
Material Borosilicate glass