EMD Millipore UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Manufacturer:  EMD Millipore ZLXUVLP01
 View more versions of this product
Catalog No. ZLXUVLP01
Specifications Description & Specifications
Specifications
Elix™ 3/5/10 water purification system
UV lamp
254nm