Please login to your online account to display your discounted pricing
EMD Millipore UV Lamps and Assemblies for Millipore Water Purification Systems
Control bacteria and oxidize organics
Manufacturer: emd millipore  ZMQUVLP01
 View more versions of this product
Catalog No. ZMQUVLP01
  • / Each
Description & Specifications
Specifications
Product Type UV lamp
Wavelength 254/185nm
For Use With (Equipment) Milli-Q™ gradient (purple door) and synthesis (orange door) model