Please login to your online account to display your discounted pricing

EMD Millipore Universal pH Indicator Papers, pH 1 to 10


Impregnatead with high-quality color

Manufacturer: emd millipore  1095260003

 View more versions of this product

Catalog No. M1095260003

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
Graduations 1.0-2.0-3.0-4.0-5.0-6.0-7.0-8.0-9.0-10.0
Quantity 3 Rolls
Shelf Life 3 to 5 Years
Storage Requirements 10° to 25°C
Product Type Universal pH Indicator Paper
pH Range 1 to 10