MilliporeSigma™ Universal pH Indicator Papers, pH 1 to 10
Impregnatead with high-quality color
Manufacturer:  MilliporeSigma™ 1095250003
 View more versions of this product
Catalog No. M1095250003
Description
Description
Color Scale: 1, 2, 3, 4, 5, 6, 7, 8, 9, 10
Specifications
Specifications
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
Each
Universal pH Indicator Paper
1.0-2.0-3.0-4.0-5.0-6.0-7.0-8.0-9.0-10.0
10° to 25°C
1 to 10