missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Universal pH Indicator Papers, pH 1 to 10

Impregnatead with high-quality color

$139.14 - $139.17

Specifications

pH Range 1 to 10
Product Type Universal pH Indicator Paper
For Use With (Application) Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
Graduations 1.0-2.0-3.0-4.0-5.0-6.0-7.0-8.0-9.0-10.0
Storage Requirements 10° to 25°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1095250003 View Documents MilliporeSigma™
1095250003
Each Each for $139.17
 
M1095260003 View Documents MilliporeSigma™
1095260003
3 Rolls Each for $139.14
 
Description

Description

Color Scale: 1, 2, 3, 4, 5, 6, 7, 8, 9, 10

Specifications

Specifications

1 to 10
Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination
10° to 25°C
Universal pH Indicator Paper
1.0-2.0-3.0-4.0-5.0-6.0-7.0-8.0-9.0-10.0