EMD Millipore Ultrapure Water for Molecular Biology
For use in critical molecular biology experiments
$46.54 - $498.16
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09-739-009 EMD Millipore
H20MB0506
500mL; 6/Pk. Pack for $236.08
09-739-008 EMD Millipore
H20MB0501
500mL; 1/Pk. Each for $46.54
09-739-007 EMD Millipore
H20MB0124
125mL; 24/Pk. Pack for $498.16
09-739-006 EMD Millipore
H20MB0106
125mL; 6/Pk. Pack for $161.20
09-739-010 EMD Millipore
H20MB1001
1L; 1/Pk. Each for $74.88
09-739-011 EMD Millipore
H20MB1006
1L; 6/Pk. Pack for $343.00
Specifications
Description
  • Sterile
  • RNase-free, DNase-free and protease-free
  • Calcium and magnesium free
  • RT-PCR Tested
  • Available in 1, 0.5 and 125mL bottles in different package sizes
Water purification; sterilizing filtration; aseptic filling and sterility testing; biochemistry; and molecular biology applications
Certifications
ISO 9001:2000/cGMP environment
Description & Specifications
Specifications
0.0