MilliporeSigma™ Ultrapure Water for Cell Culture
Cell-culture tested for use in critical cell culture experiments
$362.00 - $362.00
Chemical Identifiers
Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
09-739-001
View Documents
MilliporeSigma™
H20CC0124
24 x 125mL 24/Pk.
Pack for $362.00
Description
Description
  • Sterile
  • RNase-free, DNase-free pyrogen and protease-free
  • Calcium and magnesium free
  • Available in 1, 0.5 and 125mL bottles in different package sizes
Water purification, sterilizing filtration; aseptic filling and sterility testing; biochemistry; and cell culture
Certifications
ISO 9001:2000/cGMP environment
Specifications
Chemical Identifiers
0.0
icon-pdf.png
Specifications
24/Pk.
24 x 125mL
SDS