Please login to your online account to display your discounted pricing

EMD Millipore Ultrapure Water for Cell Culture


Cell-culture tested for use in critical cell culture experiments

Manufacturer: emd millipore  H20CC1006

 View more versions of this product

Catalog No. 09-739-005

  • / Pack
 

Description & Specifications

Specifications

Packaging 6/Pk.
Additional Information Used in cell culture
Purity Grade Ultrapure
Sterility Yes
Quantity 1L
Product Type Ultrapure Water
For Use With (Application) Cell culture
Sterile Yes
Sterile Yes
Quantity 6/Pack
Volume (Metric) 1L