missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Ultrafree™-CL Centrifugal Filter Devices with Durapore™ Membrane

Fast, convenient sample preparation; very low hold-up volumes (10μL)

Manufacturer:  EMD Millipore UFC40SV25

 View more versions of this product

Catalog No. UFC40SV25


 

Specifications

Description

Ultrafree-CL with Durapore Membrane, Non-Sterile, Dark Green, Pore Size: 5.0μm

Ideal for particle removal and purification. Ultrafree-CL processes up to 2mL.

Minimize sample handling to avoid significant losses of valuable material Low hold-up volume of sample allows highest recoveries Color coded to indicate membrane type and pore size
Polypropylene components
Silicone O-rings
No adhesives used in assembly
Low protein binding Filter insert fits standard 0.5mL microcentrifuge tubes

Description & Specifications

Specifications

Durapore
Ultrafree-CL
5/8 in.
31/32 in.
5,000xG
No
2mL
5μm
Dark Green
16.3mm
77mm
25/Pk.
0.8cm2