Please login to your online account to display your discounted pricing

EMD Millipore Ultrafree™-CL Centrifugal Filter Devices with Durapore™ Membrane


Fast, convenient sample preparation; very low hold-up volumes (10μL)

Manufacturer: emd millipore  UFC40DV25

 View more versions of this product

Catalog No. UFC40DV25

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Diameter (English) 5/8 in.
Diameter (Metric) 16.3mm
Length (English) 31/32 in.
Max. Force 5,000xG
Quantity 25/Pk.
Length (Metric) 77mm
Volume (Metric) 2mL
Color Purple
Surface Area 0.8cm2
Sterile No
Membrane Material Durapore
Pore Size 0.65μm
Type Ultrafree-CL