Please login to your online account to display your discounted pricing

EMD Millipore Ultrafree™-CL Centrifugal Filter Devices with Durapore™ Membrane


Fast, convenient sample preparation; very low hold-up volumes (10μL)

Manufacturer: emd millipore  UFC40VV00

 View more versions of this product

Catalog No. UFC40VV00

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Length (English) 31/32 in.
Length (Metric) 77mm
Max. Force 5,000xG
Quantity 100/Pk.
Sterile No
Surface Area 0.8cm2
Volume (Metric) 2mL
Membrane Material PVDF; PTFE
Pore Size 10μm
Type Ultrafree-CL
Color Orange
Diameter (English) 5/8 in.
Diameter (Metric) 16.3mm