Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLAC02510

 View more versions of this product

Catalog No. PLAC02510

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filter Material Regenerated Cellulose
For Use With (Application) Protein purification, protein concentration, protein desalting
Quantity 10 Pack
Diameter (Metric) Filter 2.5cm
Molecular Weight 1kDa
Filter Type Ultrafiltration
Product Type Ultrafiltration Disc
Filtration Area 4.1cm2
Filter Code PLAC

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules