Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLGC15005

 View more versions of this product

Catalog No. PLGC15005

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Product Type Ultrafiltration Disc
Filter Type Ultrafiltration
Quantity 5 Pack
Diameter (Metric) Filter 15cm
Filtration Area 177cm2
For Use With (Application) Protein purification, protein concentration, protein desalting
Molecular Weight 10kDa
Filter Code PLGC
Filter Material Regenerated Cellulose