Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLGC06210

 View more versions of this product

Catalog No. PLGC06210

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filtration Area 28.7cm2
Diameter (Metric) Filter 6.35cm
Filter Type Ultrafiltration
Product Type Ultrafiltration Disc
Molecular Weight 10kDa
Filter Code PLGC
For Use With (Application) Protein purification, protein concentration, protein desalting
Filter Material Regenerated Cellulose
Quantity 10 Pack