Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLAC04310**

 View more versions of this product

Catalog No. PLAC04310

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Product Type Ultrafiltration Disc
Filtration Area 13.4cm2
Diameter (Metric) Filter 4.45cm
Quantity 10 Pack
Filter Code PLAC
Filter Type Ultrafiltration
Filter Material Regenerated Cellulose
Molecular Weight 1kDa
For Use With (Application) Protein purification, protein concentration, protein desalting