Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLBC15005

 View more versions of this product

Catalog No. PLBC15005

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Product Type Ultrafiltration Disc
Filtration Area 177cm2
Filter Code PLBC
Filter Type Ultrafiltration
For Use With (Application) Protein purification, protein concentration, protein desalting
Quantity 5 Pack
Molecular Weight 3kDa
Filter Material Regenerated Cellulose
Diameter (Metric) Filter 15cm

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules