Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLBC06210

 View more versions of this product

Catalog No. PLBC06210

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10 Pack
Diameter (Metric) Filter 6.35cm
Filter Code PLBC
Filter Material Regenerated Cellulose
Filter Type Ultrafiltration
Filtration Area 28.7cm2
Molecular Weight 3kDa
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc