Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLTK15005

 View more versions of this product

Catalog No. PLTK15005

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Filter Type Ultrafiltration
Filtration Area 177cm2
Quantity 5 Pack
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc
Diameter (Metric) Filter 15cm
Molecular Weight 30kDa
Filter Code PLTK
Filter Material Regenerated Cellulose