Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLTK06210

 View more versions of this product

Catalog No. PLTK06210

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Product Type Ultrafiltration Disc
For Use With (Application) Protein purification, protein concentration, protein desalting
Filter Type Ultrafiltration
Molecular Weight 30kDa
Quantity 10 Pack
Diameter (Metric) Filter 6.35cm
Filter Code PLTK
Filter Material Regenerated Cellulose
Filtration Area 28.7cm2

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules