Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLTK04710

 View more versions of this product

Catalog No. PLTK04710

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Diameter (Metric) Filter 4.7cm
Filter Code PLTK
Filter Type Ultrafiltration
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc
Quantity 10 Pack
Filter Material Regenerated Cellulose
Molecular Weight 30kDa