Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLHK07610

 View more versions of this product

Catalog No. PLHK07610

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Product Type Ultrafiltration Disc
Filter Type Ultrafiltration
Filtration Area 41.8cm2
For Use With (Application) Protein purification, protein concentration, protein desalting
Filter Code PLHK
Diameter (Metric) Filter 7.6cm
Filter Material Regenerated Cellulose
Molecular Weight 100kDa
Quantity 10 Pack

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules