Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLHK04710

 View more versions of this product

Catalog No. PLHK04710

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filter Type Ultrafiltration
Diameter (Metric) Filter 4.7cm
Quantity 10 Pack
Molecular Weight 100kDa
Filter Material Regenerated Cellulose
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc
Filter Code PLHK