Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLHK02510

 View more versions of this product

Catalog No. PLHK02510

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filter Type Ultrafiltration
Filter Material Regenerated Cellulose
Product Type Ultrafiltration Disc
For Use With (Application) Protein purification, protein concentration, protein desalting
Diameter (Metric) Filter 2.5cm
Filter Code PLHK
Filtration Area 4.1cm2
Molecular Weight 100kDa
Quantity 10 Pack

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules