Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLGC09005

 View more versions of this product

Catalog No. PLGC09005

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Quantity 5 Pack
Diameter (Metric) Filter 9cm
Filter Code PLGC
Filter Material Regenerated Cellulose
Filter Type Ultrafiltration
Filtration Area 63cm2
Molecular Weight 10kDa
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc