Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLGC04710

 View more versions of this product

Catalog No. PLGC04710

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10 Pack
Diameter (Metric) Filter 4.7cm
Filter Material Regenerated Cellulose
Filter Code PLGC
Filter Type Ultrafiltration
Molecular Weight 10kDa
For Use With (Application) Protein purification, protein concentration, protein desalting
Product Type Ultrafiltration Disc