Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLGC02510

 View more versions of this product

Catalog No. PLGC02510

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filter Code PLGC
Diameter (Metric) Filter 2.5cm
For Use With (Application) Protein purification, protein concentration, protein desalting
Filter Type Ultrafiltration
Quantity 10 Pack
Product Type Ultrafiltration Disc
Filtration Area 4.1cm2
Filter Material Regenerated Cellulose
Molecular Weight 10kDa