missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc

Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer:  EMD Millipore PLCC07610

 View more versions of this product

Catalog No. PLCC07610


 

Specifications

Description

Ultrafiltration Disc; Millipore Ultracel; PLCC; Regenerated cellulose; 5kDA NMWL; Dia., 76mm

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules

Description & Specifications

Specifications

7.6cm
Regenerated Cellulose
41.8cm2
Protein purification, protein concentration, protein desalting
10 Pack
PLCC
Ultrafiltration
5kDa
Ultrafiltration Disc
Promotions

Promotions