Please login to your online account to display your discounted pricing

EMD Millipore Ultracel™ Ultrafiltration Disc


Use to concentrate or desalt dilute solutions with the Stirred Cell device

Manufacturer: emd millipore  PLCC04710

 View more versions of this product

Catalog No. PLCC04710

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Filter Type Ultrafiltration
For Use With (Application) Protein purification, protein concentration, protein desalting
Molecular Weight 5kDa
Diameter (Metric) Filter 4.7cm
Filter Code PLCC
Product Type Ultrafiltration Disc
Filter Material Regenerated Cellulose
Quantity 10 Pack

  • Hydrophilic with tight microstructure
  • Assures highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules