EMD Millipore Tube Adapters
Adapts 1/8 or 1/4 in. NPTM fittings to 1/4 in. OD tubing
$33.00 - $62.50
Specifications
Product Type Tube Adapter
For Use With (Equipment) Water Purification System
Connection Type NPTM to tube
Material Stainless Steel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inside Diameter (Metric) Inside Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inside Diameter (Metric) Inside Price Quantity  
XX6702507 EMD Millipore
XX6702507
0.25 in. 0.63cm Each for $62.50
YT3009018 EMD Millipore
YT3009018
0.125 in. 0.3175cm Each for $33.00
Specifications
Description
  • Made from Series 300 stainless-steel
  • Adapter (XX6702507) is a replacement part for Filterjet™ Solvent Dispenser and is included in Connector Kit for ZHE Hazardous Waste Filtration System
Description & Specifications
Specifications
Tube Adapter
Water Purification System
NPTM to tube
Stainless Steel