MilliporeSigma™ TLC Silica Gel 60 F254: 100 Glass plates, 2.5 x 7.5cm
Allows analyzation of any substance when combined with suitable mobile phase
Manufacturer:  MilliporeSigma™ 1057940001
Catalog No. M1057940001
Description
Description
Allows analyzation of any substance when combined with suitable mobile phase
  • Combination of unmodified silica gel 60 and polymeric binder for hard and adherent surface, resistant to crack and damage
  • Dense plate coating ensuring narrow bands and maximum sample separation efficiency
  • Layer thickness of 250μm with mean particle size of 10 to 12μm
  • Green fluorescencing inorganic florescent indications for UV detection of colorless substance
  • Disposable
  • Fibre case packaging
  • In-process control in drug synthesis
  • Identity and stability testing of drugs
  • Quality control of pharmaceuticals, food and environmental samples
  • Residue analysis in food and environmental samples
Specifications
Specifications
20 to 200 min.
254 nm
2.5 x 7.5 cm
30 µm
10.0
5 to 20 µm
60 Å
Classical Silica TLC plate
520m2/g
2.5 cm
Glass
In-Process Control in Drug Synthesis, Identity and Stability Testing of Drugs, Quality Control of Pharmaceuticals, Food and Environmental Samples, Residue Analysis in Food and Environmental Samples
0.0011 in.
7.5 cm
1.0
10 to 12 µm
0.8 mL/g
1/Pk.
250 µm
Silica gel 60 F254
Documents