MilliporeSigma™ TLC and HPTLC GLP Plates, Glass Backed
For thin layer chromatography applications.
$388.58 - $1,757.94
Specifications
For Use With (Application) GLP Applications
Quantity 25 Plates
View More Specs
Products 4
Catalog Number Mfr. No. Format Product Type Price Quantity  
Catalog Number Mfr. No. Format Product Type Price Quantity  
M1055660001 View Documents MilliporeSigma™
1055660001
20 x 20 cm GLP Silica TLC plate Each for $1,048.74
M1055640001 View Documents MilliporeSigma™
1055640001
10 x 10 cm HPTLC GLP silica gel 60 F254 Each for $388.58
M1057020001 View Documents MilliporeSigma™
1057020001
10 x 20 cm HPTLC GLP silica gel 60 F254 Each for $456.73
M1133260001 View Documents MilliporeSigma™
1133260001
10 x 20 cm HPTLC GLP silica gel 60 Each for $1,757.94
Description
Description
These plates feature individual laser coding for GLP applications. Designed for GLP applications, the tops of each of these plates bear an item, batch, and individual plate number. As a result, lab professionals can easily record and archive every plate they use. Based on our proven silica found in TLC and HPTLC plates, these GLP plates deliver equally unsurpassed separation performance.
Specifications
Specifications
GLP Applications
25 Plates