Please login to your online account to display your discounted pricing

EMD Millipore TLC and HPTLC GLP Plates, Glass Backed


Manufacturer: emd millipore  1133260001

 View more versions of this product

Catalog No. M1133260001

  • / Each
 

Description & Specifications

Specifications

Product Type HPTLC GLP silica gel 60
For Use With (Application) GLP applications
Quantity 25 Plates
Format 10 x 20cm