Please login to your online account to display your discounted pricing
EMD Millipore Synergy Ultrapure Water Systems Cartridges
Manufacturer: emd millipore  SIMPAK0D2
 View more versions of this product
Catalog No. SIMPAK0D2
  • / Each
Description & Specifications
Specifications
Product Type Cartridge
For Use With (Equipment) Purifies water that has previously been treated by mixed bed ion exchange (DI)
Model SimPak 2
Feedwater Source Deionized