EMD Millipore Synergy Ultrapure Water Systems Cartridges
For Simplicity Systems
$435.00 - $435.00
Specifications
Product Type Cartridge
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Feedwater Source Model For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Feedwater Source Model For Use With (Equipment) Price Quantity  
SIMPAK0D2 EMD Millipore
SIMPAK0D2
Deionized SimPak 2 Purifies water that has previously been treated by mixed bed ion exchange (DI) Each for $435.00
SIMPAK0R1 EMD Millipore
SIMPAK0R1
Elix or RiOs SimPak 1 Purifies water that has previously been treated by EDI, reverse osmosis or distillation Each for $435.00
Specifications
Description
For Simplicity® systems with DI water feed
Description & Specifications
Specifications
Cartridge