missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Synergy™ Ultrapure Water Purification System
Obtain Type 1 reagent grade water with a flow rate above 1L/min when using the Millipore Synergy™ Ultrapure Water Purification System.
Supplier: MilliporeSigma™ SYNS0HFUS
Description
Synergy systems are designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water.
- Produce Type 1 water (18.2 MOhm. cm at 25°C ultrapure water) on demand from pretreated water (reverse osmosis or demineralized water)
- Maintenance reduced to a single cartridge change once or twice a year, depending on the volume produced and the quality of the pure water fee
- Designed for users needing between 2 and 100 liters of ultrapure water per day
- Quality suitable for HPLC and other spectrophotometric/spectroscopic analytical techniques for biological, biochemical, and cell culture applications
- Dispensing adapted to different size laboratory glassware
- Time-saving fixed-volume dispense
- Compact design, along with the versatile Remote dispenser, ensure easy installation anywhere in your laboratory
- Dimensions: 15D x 21.3W x 11.4 in. H (38 x 54 x 29cm)
- Electrical feed voltage: 100-250V ±10%
- Electrical feed frequency:50-60Hz ±10%
- Synergy UV System: For applications requiring very low organic contaminant levels,it incorporates a dual wavelength UV lamp to produce water with <5ppb TOC (when feed water TOC is below 50ppb), making it suitable for HPLC, GC, ILC and TOC analyses
LC buffer preparation, HPLC isocratic and gradient solution preparation, Sample dilution, Buffer reagents for molecular biology and cell culture media preparation, Preparation of chemical solutions used with titrators, spectrophotometers and electrophoresis systems
Specifications
Pretreated | |
Type 1 | |
<10 ppb | |
4.3 psi (0.3 bar) | |
n.a. | |
29 cm | |
54 cm | |
38 cm | |
100/240 V |
1 L/min. | |
18.2 Mohm.cm product water | |
Pure to Ultrapure Purification System | |
Designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water | |
11.4 in. | |
21.3 in. | |
15 in. | |
100/240 V 50/60 Hz | |
50/60 Hz |