Please login to your online account to display your discounted pricing

EMD Millipore Synergy™ Ultrapure Water Purification System


Obtain Type 1 reagent grade water with a flow rate above 1L/min when using the Millipore Synergy™ Ultrapure Water Purification System.

Manufacturer: emd millipore  SYNSVHFUS

 View more versions of this product

Catalog No. SYNSVHFUS

  • / Each
 

Description & Specifications

Specifications

Product Type UV pure to pure and ultrapure purification system
Includes 185nm photooxidation UV lamp
Depth (Metric) 38cm
Flow Rate 1L/min.
Total Organic Carbon <5ppb
Depth (English) 15 in.
Width (English) 11.4 in.
Hertz 50/60Hz
Height (Metric) 54cm
Height (English) 21.3 in.
Electrical Requirements 100/240V 50/60Hz
For Use With (Application) Designed for laboratories that already have access to purified water and need a point-of-use system to supply high quality ultrapure water
Feedwater Source Pretreated water
Feedwater Pressure 4.3psi (0.3 bar)
Product Water Type Type 1 reagent grade
Voltage 100/240V
Width (Metric) 29cm
Resistivity 18.2 Product water