missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Sterile Syringe Filter Units

A 25mm diameter sterile syringe filter with a 0.8μm pore size Mixed Cellulose Esters membrane. Comes in a pack of 50.

$259.00 - $346.00

Specifications

Diameter (Metric) 25mm
View More Specs

Products 2
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
SLAAV255F
View Documents
MilliporeSigma™
SLAAV255F
0.8μm
Pack of 50 Each for $259.00
 
SLGSV255F
View Documents
MilliporeSigma™
SLGSV255F
0.22μm
Pack of 50 Each for $346.00
 
Description

Description

  • Used in clarification of water, buffers or aqueous solutions
  • Sterilization of drugs or aqueous solutions administered intravenously
  • Sterile syringe filter
  • Mixed cellulose ester
  • FIlter code: GS
  • Fitting inlet: Female Luer Lok
  • Fitting outlet: Male Luer slip
  • Color code: Clear inlet/blue outlet
  • Sample type: Aqueous
  • Wettablity: Hydrophillic
  • Maximum inlet pressure: 5.2 bar (75 psi)
  • Maximum operating pressure: 45°C
Specifications

Specifications

25mm
SDS