missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Sterile Syringe Filter Units

Manufacturer:  EMD Millipore SLGSV255F

 View more versions of this product

Catalog No. SLGSV255F


 

Specifications

Description

Unit, Filter; Millipore; Millex-GS; Mixed cellulose esters; With vented inlet; Ethylene oxide sterilized; Pore Size: 0.22um; Diameter: 25mm

  • Used in clarification of water, buffers or aqueous solutions
  • Sterilization of drugs or aqueous solutions administered intravenously
  • Sterile syringe filter
  • Mixed cellulose ester
  • FIlter code: GS
  • Fitting inlet: Female Luer Lok
  • Fitting outlet: Male Luer slip
  • Color code: Clear inlet/blue outlet
  • Sample type: Aqueous
  • Wettablity: Hydrophillic
  • Maximum inlet pressure: 5.2 bar (75 psi)
  • Maximum operating pressure: 45°C
Description & Specifications

Specifications

25mm
0.22μm
Promotions

Promotions