Please login to your online account to display your discounted pricing

EMD Millipore Sterifil™ 47mm Aseptic Vacuum Filter System Replacement Parts


Used for general filtration applications and filtration of samples for particulate or biological contamination analysis

Manufacturer: emd millipore  XX1104705

 View more versions of this product

Catalog No. XX1104705

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 250mL
Description 250mL Receiver Flask
For Use With (Equipment) Millipore Sterifil 47mm Aseptic Vacuum Filter System