EMD Millipore Stainless-Steel Pressure Filter Holders
Ultraclean or sterilize liquids or gases by pressure filtration
$1,335.00 - $2,763.00
Specifications
Description Filter Holder
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Height (English) Material Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Height (English) Material Price Quantity  
YY3009000 EMD Millipore
YY3009000
4.75 in. 6.75 in. PTFE Each for $1,335.00
YY3014236 EMD Millipore
YY3014236
7.25 in. 10.62 in. Stainless Steel Each for $2,763.00
Specifications
Description
  • Autoclavable with MF (mixed cellulose esters) filter in place
  • Type 316 stainless steel
  • Anodized aluminum legs
  • PTFE O-rings
90mm Filter
  • 1/4 NPTF inlet
  • Outlet with connections supplied for 0.87 in. I.D. (9.5mm) hose
  • 1/8 NPTF upstream port for vent/relief valve
142mm Filter
  • 11/8 NPTF TC flange with clamps and adapters supplied for 0.56 in. I.D. (14mm) hose
  • 1/8 NPTF upstream port for vent/relief
Other O-ring materials are available, as well as accessory cylinders to convert holders into pressure cells without separate reservoirs. Call your Fisher Scientific™ Customer Service Representative for details.
Description & Specifications
Specifications
Filter Holder