Please login to your online account to display your discounted pricing

EMD Millipore Stainless-Steel Filter Holders


For filtration of gases or liquids at inlet pressures of 20 to 700 bar

Manufacturer: emd millipore  XX4504700

 View more versions of this product

Catalog No. XX4504700

  • / Each
 

Description & Specifications

Specifications

Description Filter Holder- High Pressure
Material Stainless Steel
Diameter (Metric) Inner 47mm
Pressure 700 bar
Surface Area (Metric) 9.6cm2
Accessories

Accessories for Catalog Number: XX4504700

Item Mfr. No. Item Description Price Per Unit Quantity  
XX4504710
EMD Millipore Stainless-Steel Accessories
emd millipore
XX4504710
For filtration of gases or liquids at inlet pressures of 20 to 700 bar Pack of 5 Each for $245.00
XX4504713
EMD Millipore Stainless-Steel Accessories
emd millipore
XX4504713
For filtration of gases or liquids at inlet pressures of 20 to 700 bar Pack of 10 Each for $153.00
XX4504704
EMD Millipore Stainless-Steel Accessories
emd millipore
XX4504704
For filtration of gases or liquids at inlet pressures of 20 to 700 bar Each for $303.00
XX45047RK
EMD Millipore Stainless-Steel Accessories
emd millipore
XX45047RK
For filtration of gases or liquids at inlet pressures of 20 to 700 bar Each for $713.00
XX4504705
EMD Millipore Stainless-Steel Accessories
emd millipore
XX4504705
For filtration of gases or liquids at inlet pressures of 20 to 700 bar Pack of 5 Each for $180.00