Please login to your online account to display your discounted pricing
EMD Millipore Stainless-Steel 13 and 25mm Filter Holders
Ultraclean or sterilize small volumes dispensed by syringe
Manufacturer: emd millipore  XX3002500
 View more versions of this product
Catalog No. XX3002500
  • / Each
Description & Specifications
Specifications
Description Filter holder
Surface Area (Metric) 3.9cm2
Material Stainless Steel, PTFE
Diameter (Metric) Inner 25mm