EMD Millipore Stainless-Steel 13 and 25mm Filter Holders
Ultraclean or sterilize small volumes dispensed by syringe
$117.00 - $246.00
Specifications
Description Filter holder
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Surface Area (Metric) Diameter (Metric) Inner Material Price Quantity  
Catalog Number Mfr. No. Surface Area (Metric) Diameter (Metric) Inner Material Price Quantity  
XX3001200 EMD Millipore
XX3001200
0.8cm2 13mm Stainless Steel Each for $117.00
XX3002500 EMD Millipore
XX3002500
3.9cm2 25mm Stainless Steel, PTFE Each for $246.00
Specifications
Description
  • Stainless-steel housing
  • PTFE gaskets (13, 25mm) and O-ring (13mm)
  • Autoclavable with filter in place
  • Filtration area: 13mm Swinnex™ style, 0.8cm2; 25mm Microsyringe style, 3.9cm2
Description & Specifications
Specifications
Filter holder